Order Entry
Ireland
ContactUsLinkComponent
1,1,2-Trimethyl-3-(4-sulphobutyl)benz[e]benzindolium inner salt 90% (dye content), Sigma-Aldrich®
1,1,2-Trimethyl-3-(4-sulphobutyl)benz[e]benzindolium inner salt 90% (dye content), Sigma-Aldrich®
Catalog # 646679-5G
CAS Number:  
undefined
1,1,2-Trimethyl-3-(4-sulphobutyl)benz[e]benzindolium inner salt 90% (dye content), Sigma-Aldrich®
Catalog # 646679-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :345.46 g/mol
Melting Pt:245…258 °C
Storage Temperature:Ambient
CAS Number:63149-24-6
Formula:C₁₉H₂₃NO₃S
MDL Number:MFCD02690163