Ethyl viologen diiodide 99%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1983-61-5
:
Formula:C₁₄H₁₈I₂N₂
MDL Number:MFCD00799539
:468.12 g/mol
Melting Pt:275 °C (dec., lit.)
:M-Clarity™ quality level = MQ100
Specification Test Results
- Appearance:Orange to red powder, crystals, crystalline Powder and/or chunks
- Infrared spectrum:Conforms to structure
- Elemental analysis:Carbon 35.4%-36.5%, Nitrogen 5.8%- 6.2%
- Titration:98.5%-101.5% (with silver nitrate)
- High pressure liquid:98.5% (minimum)
Specification Test Results
Frequently Bought Together