Order Entry
Ireland
ContactUsLinkComponent
 
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97% , Sigma-Aldrich®
 
undefined
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 97% , Sigma-Aldrich®

 

  • Appearance (Colour):
    Red to Very Dark Red and Purple and Red-Purple and Purple-Red
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    49.7 - 53.4%
  • Nitrogen:
    15.4 - 16.6%
  • Purity (HPLC):
    ≥96.5%

 

Frequently Bought Together