Order Entry
Ireland
ContactUsLinkComponent
 
 
 
4-Nitrobenzenediazonium tetrafluoroborate 97%, Sigma-Aldrich®
  294438-10G
 :  
undefined
 
 
4-Nitrobenzenediazonium tetrafluoroborate 97%, Sigma-Aldrich®
  294438-10G
 :  

 

  • Pk:
    10 g
  • Seq:
    3236

 

 

 :
 :236.92 g/mol
 :144,..148 °C (dec.)
 :1,66 g/cm³ (25 °C)
 :Refrigerator
 :207-261-1
 :8,2
 :GHS05
 :GHS07
 :456-27-9
 :C₆H₄BF₄N₃O₂
 :MFCD00012005
 :8,II