Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Benzylidenebenzenesulphonamide
N-Benzylidenebenzenesulphonamide ≥97%, Sigma-Aldrich®
N-Benzylidenebenzenesulphonamide ≥97%, Sigma-Aldrich®
301108-5G
:
SIGMA ALDRICH MICROSCOPY
:
N-Benzylidenebenzenesulphonamide ≥97%, Sigma-Aldrich®
301108-5G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
5 g
Seq:
3173
:
:
245.30 g/mol
:
78…81 °C (lit.)
:
GHS07
:
Ambient
:
13909-34-7
:
C₁₃H₁₁NO₂S
:
MFCD00011587