Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:3520-42-1
:
Formula:C₂₇H₃₀N₂O₇S₂
MDL Number:MFCD00010180
:580.65 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
EINECS:222-529-8
Specification Test Results
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Wavelength, In Methanolic Sodium Hydroxide:554 - 560 nm
- Extinction Coefficient:≥85000
- Carbon:≥40.8 %
- Nitrogen:≥3.5 %
- Dye Content:≥72 %
- Recommended Retest Period:5 Years
Specification Test Results
Frequently Bought Together