Zinquin ethyl ester ≥95% (by HPLC), Sigma-Aldrich®
Catalog # Z2251-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester
:
:414.47 g/mol
Storage Temperature:Refrigerator
CAS Number:181530-09-6
Formula:C₂₁H₂₂N₂O₅S