Haematoxylin (Harris), Sigma-Aldrich®
Catalog # HHS16-500ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:302.28 g/mol
Storage Temperature:Ambient
:1
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111