EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], powder, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:70539-42-3
:
Formula:C₁₈H₂₀N₂O₁₂
MDL Number:MFCD00049057
:456.36 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
Specification Test Results
- Appearance (Colour):White to Off-White
- Appearance (Form):Powder
- Solubility (Colour):Colorless to Faint Yellow
- Solubility (Turbidity)
50 mg/mL in HOAc:H2O (1:1):Clear to Hazy - Proton NMR spectra:Conforms to Structure
- 13C NMR SPECTRA:Conforms to Structure
- Carbon:46.1 - 48.6 %
- Nitrogen:5.8 - 6.6 %
Specification Test Results
Frequently Bought Together