Fast Blue RR Salt, Sigma-Aldrich®
Catalog # F0500-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:387.89 g/mol
Storage Temperature:Freezer
CAS Number:14726-29-5
Formula:C₁₅H₁₄ClN₃O₃
MDL Number:MFCD00074762