Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fast Red Violet LB Base solution
Fast Red Violet LB Base solution, Sigma-Aldrich®
Fast Red Violet LB Base solution, Sigma-Aldrich®
Fast Red Violet LB Base solution, Sigma-Aldrich®
912-10ML
:
SIGMA ALDRICH MICROSCOPY
:
Fast Red Violet LB Base solution, Sigma-Aldrich®
912-10ML
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
10 mL
Seq:
2275
:
:
8,III
:
GHS05
:
Refrigerator
:
1
:
MFCD00007780