Clozapine
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine
CAS Number:5786-21-0
:
Formula:C₁₈H₁₉ClN₄
MDL Number:MFCD00153785
Molecular Weight:326.83 g/mol
Melting Pt:182…185 °C
UN:2811
ADR:6.1 III
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:183 to 187 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together