Order Entry
Ireland
ContactUsLinkComponent
 
Tinidazole
 
undefined
Tinidazole

 

  • Appearance:
    Almost white to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    125 to 128 °C
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥97.5 %

 

Frequently Bought Together