Order Entry
Ireland
ContactUsLinkComponent
β-Cyclodextrin sulphobutyl ethers sodium salts
Irritant
β-Cyclodextrin sulphobutyl ethers sodium salts
Certificates
undefined
Irritant
β-Cyclodextrin sulphobutyl ethers sodium salts

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤10 %
  • Solubility:
    (30% in water) Clear solution
  • HPLC:
    ≥97.5 %
  • pH:
    4.0 to 6.8 (30% in water)

Specification Test Results

Frequently Bought Together