Fluvoxamine maleate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61718-82-9
:
Formula:C₁₅H₂₁F₃N₂O₂
MDL Number:MFCD00269809
:434.41 g/mol
Melting Pt:120…122 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:120 to 124 °C
- Solubility:(5 % in ethanol) Clear solution
- Residue after ignition:≤0.1 %
- Titration with HClO4:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together