1-Bromonaphthalene ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:90-11-9
:
Formula:C₁₀H₇Br
MDL Number:MFCD00003868
:207.07 g/mol
EINECS:201-965-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:≥ 96 %
- Appearance:Clear yellowish oily liquid:Passes test
- Density (20/4):1,483 - 1,486
Specification Test Results
Frequently Bought Together