Order Entry
Ireland
ContactUsLinkComponent
Cleanroom wipes, pre-saturated, alcohol, Kimtech™ W4
  KIMB06070
undefined
Cleanroom wipes, pre-saturated, alcohol, Kimtech™ W4
  KIMB06070
  • Packed:
    400 wipes/case
  • L×W:
    228,6×279,4 mm
  • Pk:
    400
The 100% polypropylene wipers come pre-saturated with 70% isopropyl alcohol (semiconductor grade) and 30% deionized water, in a resealable pouch.
  • Ideal one-step application of alcohol to critical surfaces
  • Low lint for environments where 'lint free' standards are important
  • These disposable wipers are recommended for ISO Class 4 and higher clean rooms
These disposable wipers are ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries that require alcohol wiping for critical tasks.
 : Meets EPA guidelines for minimum post consumer waste content.
 : Package containe 40 wipes/pack, 10 packs/case, 400 wipes/case.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904