- CE-IVD:+
- Colour code:Blue
- Packed:5 racks, 96 each
- Sterile:+
- Volume:50 - 1000 µl
- Length:71 mm
- Seq:04
- Tip style:Biopur®
- Pk:480
Tip to tip premium quality ensures reproducible results with epT.I.P.S. pipette tips in system with Eppendorf pipettes. Specific needs for your application are satified by a variety of purities, volumes and tip lengths.
- Biopur pipette tips offer the highest biological purity grade
- Certified PCR clean, sterile, pyrogen-, ATP- and DNA-free
- For use in medical, pharmaceutical and food industry, molecular biology and cell technology
- Each production lot inspected by an independent laboratory
- Also pre-sterilised ep Dualfilter T.I.P.S. SealMax are offered in racks
- Modern slim rack design with reclosable lid
epT.I.P.S. racks - Biopur pipette tips - maximum biological purity, meet most stringent requirements of the medical, pharmaceutical, food industry and of molecular biology and cell biology. Lot no and expiration date on each rack label, lot-specific certificates by independent laboratory online.
These pipette tips are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned epT.I.P.S.® Racks are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.