Order Entry
Ireland
ContactUsLinkComponent
Water, PESTINORM® for headspace gas chromatography
Water, PESTINORM® for headspace gas chromatography
Catalog # 88251.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Water, PESTINORM® for headspace gas chromatography
Catalog # 88251.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Color (APHA, Pt-Co):
    max. 5
  • Acidity (as Acetic acid):
    max. 0.0005 %
  • Alkalinity (as Ammonia):
    max. 0.0001 %
  • GC-HS (Each class 1 solvent):
    max. 1 ug/g
  • GC-HS (Each class 2 solvent):
    max. 10 ug/g
  • GC-HS (Each class 3 solvent):
    max. 50 ug/g
  • Headspace test for O.V.I's:
    Complying

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332