Order Entry
Ireland
ContactUsLinkComponent
 
Chlorophenol red, AnalaR NORMAPUR® indicator
 
undefined
Chlorophenol red, AnalaR NORMAPUR® indicator
 

 

  • Environmentally Preferable:

 

 

  • Green to red-brown to brown powder:
    Passes test
  • Solubility in ethanol (0.1 %):
    Clear solution
  • Visual transition interval :
    pH 4.8 - pH 6.4
  • Loss on drying:
    ≤ 8.0 %
  • Absorption max. λ (0.1 mol/l NaOH):
    572 - 575 nm
  • Absorptivity (1 %; 1 cm; λmax):
    ≥ 1000

 

Frequently Bought Together