Shoe covers, BizTex® Microporous type PB[6]
Catalog # PRTWST44WHR
Supplier: Portwest
Specifications
- Colour:White
- Pk:200
Specifications
About this item
These protective overshoes are designed for use in all work environments where shoes or floors need protecting. Elastic finish to top of shoe ensures a secure fit.
- Made of microporous polypropylene/polyethylene
Certifications: This fabric conforms to EN 14126 biological protection and EN 1149-1 anti-static protection, EN 13034 Type PB [6].
Caution: Do not iron and wash.