Safety boots, wellingtons, Portwest™ Pro FD95
Supplier: Portwest
Certificates
About this item
Antistatic boots made from more flexible and longer lasting than PVC or Rubber, due to its air bubble structure which provides air circulation. PU keeps feet hot in winter and cool in summer.
- Protective steel toecap and steel midsole
- Energy absorbing seat region
- 100% waterproof keeping feet warm and dry
- Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
- Fuel and oil resistant outsole
- Cold insulation
- Puncture resistant non-metallic midsole (PR)
This range is designed to protect your feet in the toughest environments.
Certifications: EN ISO 20345:2011, S5, SRC.
Frequently Bought Together