Gas filters, P906
Catalog # PRTWP906BKR
Supplier: Portwest
Specifications
- Description:A2 Gas filter, P906
- Pk:6
Specifications
About this item
P906 filters are anti-gas filter that protects against contaminant gases. Equipped with a standard threaded connection conforming to EN 148-1 and can be used on full face masks equipped with the same standard connection.
- Lightweight and comfortable
- Individually packed for vending machines
These gas filter protects from organic gases and vapours where the boiling point is higher than 65 °C. Classified as a class 2 filter, they can be used to protect against harmful substances, solvents and gases.
Certifications: EN 148 -1, EN 14387 Type A Class 2