Occupational shoes, slip-on, FW58
: Portwest
Washable upper with a self-cleaning outsole makes this slip on shoe ideal when a protective toecap is not required. Lightweight and comfortable construction.
- Anti-static footwear
- Energy absorbing seat region
- Water resistant upper to prevent water penetration
- Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
- Fuel and oil resistant outsole
- Mono density sole unit
Material upper: Microfibre
Material sole: PU
These shoes do not utilise animal product or biproducts.
: CE certified, EN ISO 20347 O2
Frequently Bought Together