Order Entry
Ireland
ContactUsLinkComponent
Analytical standard, Pyraclostrobin, reference material, Fluka™
Analytical standard, Pyraclostrobin, reference material, Fluka™
Catalog # HONCR1734-100MG
CAS Number:  
Analytical standard, Pyraclostrobin, reference material, Fluka™
Catalog # HONCR1734-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    175013-18-0
  • Description:
    Pyraclostrobin
  • Description:
    Pyraclostrobin
  • Form:
    Neat

Specifications

About this item