Lanthanum for AAS, (CRMs), Fluka™
Catalog # HONCE4400-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:2%
- Matrix:2% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Lanthanum
- Symbol:La
Specifications
About this item
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).