Order Entry
Ireland
ContactUsLinkComponent
 
Environmental standard for AA and ICP-OES
Catalog # HONCE2154-100ML
CAS Number:  
Environmental standard for AA and ICP-OES
Catalog # HONCE2154-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Environmental Standard

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L Mercury (Hg) in HNO₃ 5%