Order Entry
Ireland
ContactUsLinkComponent
Signs, floor signs, ToughStripe™, footprints
Signs, floor signs, ToughStripe™, footprints
Catalog # 116-0924
Supplier:  Brady
undefined
Signs, floor signs, ToughStripe™, footprints
Catalog # 116-0924
Supplier:  Brady

Specifications

  • Description:
    Floor marking footprints, ToughStripe floor marking, blue
  • Dimensions:
    254H×88,90W mm
  • Material:
    Polyester
  • Pk:
    10

Specifications

About this item

ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.

  • Floor marking products are made with B-514 polyester
  • Recommended for indoor use and dry environment
  • Resistant to harsh chemicals

0,203 mm thickness and service temperature range of ⁻18 °C to 54 °C.

Certifications: English XL Variable Incidence Tribometer per ASTM / ANSI F1679. ANSI A1264.2­2006 and OSHA 1910.22
Packaging: Footprints has 5 left and 5 right.