Thermal transfer ribbon for printer, R6400 series
: Brady
Ultra high chemical resistance at high temperatures. Superior solvent resistance.
- Core - diameter (mm): 25.40 mm
- Ribbon formulation: resin
Frequently Bought Together
Ultra high chemical resistance at high temperatures. Superior solvent resistance.