4-(Dimethylamino)pyridine ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1122-58-3
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00006418
:122.17 g/mol
Boiling Pt:195 °C (1013 hPa)
Melting Pt:109…111 °C
Flash Pt:124 °C
Storage Temperature:Ambient
EINECS:214-353-5
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Assay:Min. 99 %
- Melting point:111 --> 114 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87080.130 spec_VWRC87080.130 spec_VWRC87080.180 spec_VWRC87080.230
Product specification
Frequently Bought Together