Polyethylene glycol 200, GPR RECTAPUR®
Catalog # 87081.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Density (20/4):1.124 --> 1.126
- Hydroxyl value:535 --> 590
Specification Test Results
About this item
:
:200 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:–55…–40 °C
Density:1,125 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779596