Order Entry
Ireland
ContactUsLinkComponent
γ-Butyrolactone ≥99%, GPR RECTAPUR®
γ-Butyrolactone ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
γ-Butyrolactone ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Density (20/4):
    1.128 --> 1.129
  • Refractive index (20°C; 589 nm):
    1.435 --> 1.436
  • Water:
    Max. 0.5 %

Specification Test Results

Frequently Bought Together