Methylene blue
Product Details & Documents
Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
- CAS Number:61-73-4
- Formula:C₁₆H₁₈ClN₃S
- MDL Number:MFCD00150008
- Molecular Weight:319.86 g/mol
- Merck Index:12,06137
- Melting Pt:180 °C
- Density:1,757 g/cm³ (22 °C)
- Storage Temperature:Ambient
- EINECS:200-515-2



