Dansyl chloride 98%
:605-65-2
:
:C₁₂H₁₂ClNO₂S
:MFCD00003985
:269.75 g/mol
:13,02840
:71…74 °C
:Refrigerator
:210-092-6
:3261
:8,II
:GHS07
:GHS05
- Appearance:Yellow to light orange Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:70 to 74 °C
- HPLC:≥97.5 %
- UV:at 420 nm A: ≤0.01
- Color scale:≤15 APHA
Frequently Bought Together