Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl sulphoxide
Dimethyl sulphoxide
Dimethyl sulphoxide
Dimethyl sulphoxide
PIER20688
:
Thermo Fisher Scientific
:
Dimethyl sulphoxide
PIER20688
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle
Pk:
950 ml
Seq:
2845
:
:
78.14 g/mol
:
13,03285
:
189 °C (1013 hPa)
:
18,5 °C
:
1,101 g/cm³ (20 °C)
:
87 °C
:
Ambient
:
200-664-3
:
67-68-5
:
(CH₃)₂SO
:
MFCD00002089