

Dodecyltrimethylammonium bromide 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1119-94-4
:
Formula:C₁₅H₃₄BrN
MDL Number:MFCD00011767
:308.35 g/mol
Melting Pt:246 °C (dec.)
Storage Temperature:Ambient
EINECS:214-290-3
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- Solubility:(10 % in water) Clear to slightly hazy
- Color of solution:≤100 APHA (10% aq. soln.)
Specification Test Results
Frequently Bought Together