Sand, TECHNICAL, industrial, washed
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
MDL Number:MFCD02100519
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:2,66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:238-878-4
Specification Test Results
- Solubility in acid:Max. 1 %
Specification Test Results
Related Information
Frequently Bought Together