Strontium titanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12060-59-2
:
Formula:O₃SrTi
MDL Number:MFCD00049554
:183.5 g/mol
Melting Pt:2080 °C
Density:5,11 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:235-044-1
Frequently Bought Together