Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Strontium titanate
Strontium titanate substrate 5x5x0.5 mm, polished one side 110 orientation
Strontium titanate substrate 5x5x0.5 mm, polished one side 110 orientation
42619.KT
:
Thermo Fisher Scientific
:
Strontium titanate substrate 5x5x0.5 mm, polished one side 110 orientation
42619.KT
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
:
183.5 g/mol
:
2080 °C
:
5,11 g/cm³ (20 °C)
:
Ambient
:
235-044-1
:
12060-59-2
:
O₃SrTi
:
MFCD00049554