Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium titanate
Strontium titanate substrate 5x5x0.5 mm, polished both sides 111 orientation
Strontium titanate substrate 5x5x0.5 mm, polished both sides 111 orientation
Strontium titanate substrate 5x5x0.5 mm, polished both sides 111 orientation
42594.KT
:
Thermo Fisher Scientific
:
Strontium titanate substrate 5x5x0.5 mm, polished both sides 111 orientation
42594.KT
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
:
183.5 g/mol
:
2080 °C
:
5,11 g/cm³ (20 °C)
:
Ambient
:
235-044-1
:
12060-59-2
:
O₃SrTi
:
MFCD00049554