Strontium titanate substrate 10x10x0.5 mm, polished one side 110 orientation
Catalog # 40276.KT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:183.5 g/mol
Melting Pt:2080 °C
Density:5,11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-044-1
CAS Number:12060-59-2
Formula:O₃SrTi
MDL Number:MFCD00049554