Tungstic(VI) acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-03-1
:
Formula:H₂O₄W
MDL Number:MFCD00011336
Merck Index:14,9818
Boiling Pt:1473 °C
Melting Pt:100 °C (dec.)
Density:5,5
:249.85 g/mol
EINECS:231-975-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together