Phenazone, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:60-80-0
:
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD00003146
:188.23 g/mol
Merck Index:12,00757
Boiling Pt:388 °C (1013 hPa)
Melting Pt:152…154 °C
Density:1,25 g/cm³ (20 °C)
EINECS:200-486-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together