Disoprofol
250 mg
Catalog # PROOMM0461.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Propofol, 1,3-Di(methylethyl)-2-hydroxybenzene, 2,6-Diisopropylphenol, 2,6-Bisisopropylphenol
- CAS Number:2078-54-8
- Formula:C₁₂H₁₈O
- MDL Number:MFCD00008885
- Molecular Weight:178.27
- Merck Index:13,07927
- Boiling Pt:256 °C (1013 hPa)
- Melting Pt:18 °C
- Density:1,032 g/cm³ (20 °C)
- :Density: 1.032 g/cm³ (20 °C)
- EINECS:218-206-6
Specifications
Specifications
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



