Order Entry
Ireland
ContactUsLinkComponent
Dodecyl-3,4,5-trihydroxybenzoate for synthesis, Sigma-Aldrich®
Dodecyl-3,4,5-trihydroxybenzoate for synthesis, Sigma-Aldrich®
Catalog # 8.20598.0100
Supplier:  Merck
CAS Number:  
Dodecyl-3,4,5-trihydroxybenzoate for synthesis, Sigma-Aldrich®
Catalog # 8.20598.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    94 - 97 °C
  • Molar Mass:
    338.44 g/mol

Specification Test Results

About this item

 :
 :338.44 g/mol
Melting Pt:94…97 °C
 :M-Clarity™ quality level = MQ200
EINECS:214-620-6
 :GHS07
CAS Number:1166-52-5
Formula:C₁₉H₃₀O₅
MDL Number:MFCD00002195