Order Entry
Ireland
ContactUsLinkComponent
 
 
Sodium diatrizoate hydrate ≥98%, Sigma-Aldrich®
 
undefined
 
Sodium diatrizoate hydrate ≥98%, Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Off White
  • Appearance (Form):
    Powder
  • 1H NMR Spectrum:
    Conforms to Structure
  • Water (by Karl Fischer):
    ≤11.0 %
  • Titration
    Anhydrous:
    ≥98.0 %
  • Solubility (Colour):
    Colorless to Faint Yellow
  • Solubility (Turbidity)
    350 mg/ml, H2O:
    Clear to Slightly Hazy

 

Frequently Bought Together