MOPS-Na 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
- CAS Number:71119-22-7
- Formula:C₇H₁₄NNaO₄S
- MDL Number:MFCD00064350
- Molecular Weight:231.25 g/mol
- Storage Temperature:Ambient
- EINECS:428-420-3
3 Options Available Below
Product Details & Documents
Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
- CAS Number:71119-22-7
- Formula:C₇H₁₄NNaO₄S
- MDL Number:MFCD00064350
- Molecular Weight:231.25 g/mol
- Storage Temperature:Ambient
- EINECS:428-420-3
Documents
ACRO35259
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(23.125 g/l in cold water) Clear and colorless
- Titration with HClO4:≥97.5 %



