1-Pentanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:22767-49-3
:
Formula:C₅H₁₁O₃SNa
MDL Number:MFCD00007541
:174.2 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:245-208-4
Specification Test Results
- Assay (calculated on dried substance):≥ 99,0 %
- Appearance:White powder
- IR Spectrum:Passes test
- Water:≤ 2,0 %
- Absorbance (200 nm) (0.25 mol/l):≤ 0,1
- Absorbance (210 nm) (0.25 mol/l):≤ 0,05
- Absorbance (220 nm) (0.25 mol/l):≤ 0,04
- Absorbance (230 nm) (0.25 mol/l):≤ 0,03
- Absorbance (240 nm) (0.25 mol/l):≤ 0,01
- Absorbance (250 nm) (0.25 mol/l):≤ 0,01
- Absorbance (260 nm) (0.25 mol/l):≤ 0,01
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC152812V spec_VWRC152812V spec_VWRC152813W spec_VWRC152813W
Product specification
Frequently Bought Together