1-Cetylpyridinium chloride monohydrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6004-24-6
:
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977
:358.01 g/mol
Merck Index:13,02041
Melting Pt:77…83 °C
Storage Temperature:Ambient
EINECS:204-593-9
UN:2811
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS09
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:217 °C
- Molar Mass:358.01 g/mol
- Bulk Density:370 kg/m3
- pH:5.0 - 5.4 (10 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together