


Cobalt(II) perchlorate hexahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13478-33-6
:
Formula:Cl₂CoO₈·6H₂O
MDL Number:MFCD00150207
:365.93 g/mol
Storage Temperature:Ambient
EINECS:236-653-5
UN:1481
ADR:5.1,II
:GHS03
:GHS08
:GHS07
Related Information
Frequently Bought Together